ADVH49B82423 AChemBlock

tert-butyl 2-ethynylpiperazine-1-carboxylate

  • Cas Number : 1259014-64-6
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-ethynylpiperazine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81613
Availability details for
Quantity
  • Availability
  • Add to Cart