ADVH49B82428 AChemBlock

1-tert-Butyl-1,5,6,7-tetrahydro-pyrazolo[4,3-c]pyridin-4-one

  • Cas Number : 1257403-88-5
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-1-5-6-7-tetrahydro-pyrazolo-4-3-c-pyridin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86627
Availability details for
Quantity
  • Availability
  • Add to Cart