ADVH49B8242B AChemBlock

methyl 4-chloro-6-fluoropyridine-2-carboxylate

  • Cas Number : 1256810-49-7
  • Molecular Weight : 189.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-chloro-6-fluoropyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81661
Availability details for
Quantity
  • Availability
  • Add to Cart