ADVH49B82432 AChemBlock

rel-(3R,4R)-4-fluoropyrrolidin-3-amine;dihydrochloride

  • Cas Number : 125197-40-2
  • Molecular Weight : 177.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/rel-3r-4r-4-fluoropyrrolidin-3-amine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81741
Availability details for
Quantity
  • Availability
  • Add to Cart