ADVH49B82433 AChemBlock

7-bromoindan-1-one

  • Cas Number : 125114-77-4
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromoindan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81765
Availability details for
Quantity
  • Availability
  • Add to Cart