ADVH49B82435 AChemBlock

3-cyclohexyl-1H-pyrazol-4-amine

  • Cas Number : 1249721-12-7
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-cyclohexyl-1h-pyrazol-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81835
Availability details for
Quantity
  • Availability
  • Add to Cart