ADVH49B82436 AChemBlock

2-chloro-4-(furan-2-yl)pyrimidine

  • Cas Number : 124959-28-0
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-4-furan-2-yl-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86524
Availability details for
Quantity
  • Availability
  • Add to Cart