ADVH49B8243B AChemBlock

methyl 5-bromo-2-chloro-3-nitro-benzoate

  • Cas Number : 124371-59-1
  • Molecular Weight : 294.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-bromo-2-chloro-3-nitro-benzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S81936
Availability details for
Quantity
  • Availability
  • Add to Cart