ADVH49B82440 AChemBlock

tert-butyl 4-(hydroxymethyl)benzylcarbamate

  • Cas Number : 123986-64-1
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-hydroxymethyl-benzylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71774
Availability details for
Quantity
  • Availability
  • Add to Cart