ADVH49B82446 AChemBlock

(2S,3R)-1-tert-butoxycarbonyl-3-phenyl-pyrrolidine-2-carboxylic acid

  • Cas Number : 123724-21-0
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2s-3r-1-tert-butoxycarbonyl-3-phenyl-pyrrolidine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82021
Availability details for
Quantity
  • Availability
  • Add to Cart