ADVH49B82447 AChemBlock

3-benzylazetidin-3-ol

  • Cas Number : 1236862-03-5
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-benzylazetidin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82022
Availability details for
Quantity
  • Availability
  • Add to Cart