ADVH49B8244A AChemBlock

ABT-639

  • Cas Number : 1235560-28-7
  • Molecular Weight : 455.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/abt-639.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72383
Availability details for
Quantity
  • Availability
  • Add to Cart