ADVH49B8244D AChemBlock

N-Boc-L-glutamate 5-(9-fluorenylmethyl)ester

  • Cas Number : 123417-18-5
  • Molecular Weight : 425.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-boc-l-glutamate-5-9-fluorenylmethyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70388
Availability details for
Quantity
  • Availability
  • Add to Cart