ADVH49B8244E AChemBlock

tert-butyl 4-phenylpiperidine-1-carboxylate

  • Cas Number : 123387-49-5
  • Molecular Weight : 261.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-phenylpiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71775
Availability details for
Quantity
  • Availability
  • Add to Cart