ADVH49B82452 AChemBlock

tert-butyl 1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate

  • Cas Number : 1228079-29-5
  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-oxo-1-3-dihydrospiro-indene-2-4-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82134
Availability details for
Quantity
  • Availability
  • Add to Cart