ADVH49B8245B AChemBlock

5-methoxy-1H-indazol-6-ol

  • Cas Number : 1227270-78-1
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-1h-indazol-6-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82168
Availability details for
Quantity
  • Availability
  • Add to Cart