ADVH49B8245C AChemBlock

tert-butyl 2-chloro-5-oxo-5,6,7,8-tetrahydro-1,6-naphthyridine-6-carboxylate

  • Cas Number : 1226898-92-5
  • Molecular Weight : 282.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-chloro-5-oxo-5-6-7-8-tetrahydro-1-6-naphthyridine-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82181
Availability details for
Quantity
  • Availability
  • Add to Cart