ADVH49B8245D AChemBlock

FLLL32

  • Cas Number : 1226895-15-3
  • Molecular Weight : 464.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/flll32.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86778
Availability details for
Quantity
  • Availability
  • Add to Cart