ADVH49B82463 AChemBlock

E6446

  • Cas Number : 1219925-73-1
  • Molecular Weight : 449.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/e6446.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86675
Availability details for
Quantity
  • Availability
  • Add to Cart