ADVH49B82468 AChemBlock

piperidine-4-boronic acid pinacol ester hydrochloride

  • Cas Number : 1218790-99-8
  • Molecular Weight : 265.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/piperidine-4-boronic-acid-pinacol-ester-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82277
Availability details for
Quantity
  • Availability
  • Add to Cart