ADVH49B8246B AChemBlock

3-Bromo-5-hydroxyphenylboronic acid pinacol ester

  • Cas Number : 1218789-50-4
  • Molecular Weight : 316.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-hydroxyphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70798
Availability details for
Quantity
  • Availability
  • Add to Cart