ADVH49B8246C AChemBlock

CAY10505

  • Cas Number : 1218777-13-9
  • Molecular Weight : 289.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/cay10505.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86693
Availability details for
Quantity
  • Availability
  • Add to Cart