ADVH49B8246D AChemBlock

(2R,4S)-4-Boc-amino-pyrrolidin-2-carboxylic acid

  • Cas Number : 1217651-50-7
  • Molecular Weight : 230.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2r-4s-4-boc-amino-pyrrolidin-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70391
Availability details for
Quantity
  • Availability
  • Add to Cart