ADVH49B8246F AChemBlock

6-bromo-8-methyl-[1,2,4]triazolo[4,3-a]pyridine

  • Cas Number : 1216301-85-7
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-8-methyl-1-2-4-triazolo-4-3-a-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82334
Availability details for
Quantity
  • Availability
  • Add to Cart