ADVH49B8247F AChemBlock

7-bromo-1-methyl-3,4-dihydro-2H-pyrido[2,3-b]pyrazine

  • Cas Number : 1211581-93-9
  • Molecular Weight : 228.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-1-methyl-3-4-dihydro-2h-pyrido-2-3-b-pyrazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82411
Availability details for
Quantity
  • Availability
  • Add to Cart