ADVH49B82481 AChemBlock

6-chloro-4-(trifluoromethyl)pyridin-3-ol

  • Cas Number : 1211578-60-7
  • Molecular Weight : 197.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-trifluoromethyl-pyridin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82414
Availability details for
Quantity
  • Availability
  • Add to Cart