ADVH49B82482 AChemBlock

2-bromo-3-fluoropyridine-4-carboxylic acid

  • Cas Number : 1211530-89-0
  • Molecular Weight : 220

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-3-fluoropyridine-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71783
Availability details for
Quantity
  • Availability
  • Add to Cart