ADVH49B82483 AChemBlock

4,6-dichloro-3-methyl-1h-pyrazolo[3,4-d]pyrimidine

  • Cas Number : 1211522-68-7
  • Molecular Weight : 203.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-3-methyl-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82435
Availability details for
Quantity
  • Availability
  • Add to Cart