ADVH49B82486 AChemBlock

5-bromo-11-phenyl-11H-benzo[a]carbazole

  • Cas Number : 1210469-09-2
  • Molecular Weight : 372.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-11-phenyl-11h-benzo-a-carbazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86519
Availability details for
Quantity
  • Availability
  • Add to Cart