ADVH49B82488 AChemBlock

ethyl (1R,3R,6S)-7-oxabicyclo[4.1.0]heptane-3-carboxylate

  • Cas Number : 1210348-12-1
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1r-3r-6s-7-oxabicyclo-4-1-0-heptane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82455
Availability details for
Quantity
  • Availability
  • Add to Cart