ADVH49B8248D AChemBlock

4-chloro-7-fluoro-2-methylquinazoline

  • Cas Number : 1206694-32-7
  • Molecular Weight : 196.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-7-fluoro-2-methylquinazoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86566
Availability details for
Quantity
  • Availability
  • Add to Cart