ADVH49B8248E AChemBlock

(3S)-1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidin-3-ol

  • Cas Number : 1206641-40-8
  • Molecular Weight : 303.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3s-1-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-phenyl-methyl-pyrrolidin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82513
Availability details for
Quantity
  • Availability
  • Add to Cart