ADVH49B8248F AChemBlock

4,6-dibromopyridine-2-carbonitrile

  • Cas Number : 1206247-80-4
  • Molecular Weight : 261.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-6-dibromopyridine-2-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82521
Availability details for
Quantity
  • Availability
  • Add to Cart