ADVH49B82490 AChemBlock

6-chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • Cas Number : 1204926-24-8
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloropyrazolo-1-5-a-pyrimidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82530
Availability details for
Quantity
  • Availability
  • Add to Cart