ADVH49B82496 AChemBlock

6-chloro-3H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one

  • Cas Number : 1198475-34-1
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-3h-4h-pyrrolo-2-1-f-1-2-4-triazin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82581
Availability details for
Quantity
  • Availability
  • Add to Cart