ADVH49B8249A AChemBlock

4,6-dimethylpyridin-3-amine

  • Cas Number : 1193-71-1
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-6-dimethylpyridin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71794
Availability details for
Quantity
  • Availability
  • Add to Cart