ADVH49B8249B AChemBlock

6'-bromo-1',2'-dihydrospiro[oxane-4,3'-pyrrolo[3,2-b]pyridine]-2'-one

  • Cas Number : 1190862-19-1
  • Molecular Weight : 283.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-1-2-dihydrospiro-oxane-4-3-pyrrolo-3-2-b-pyridine-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82671
Availability details for
Quantity
  • Availability
  • Add to Cart