ADVH49B8249C AChemBlock

6-bromo-1,3,3-trimethyl-2,3-dihydro-1H-indol-2-one

  • Cas Number : 1190861-69-8
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-1-3-3-trimethyl-2-3-dihydro-1h-indol-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82672
Availability details for
Quantity
  • Availability
  • Add to Cart