ADVH49B8249D AChemBlock

5-Cyano-3-nitro-7-azaindole

  • Cas Number : 1190322-60-1
  • Molecular Weight : 188.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-cyano-3-nitro-7-azaindole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72393
Availability details for
Quantity
  • Availability
  • Add to Cart