ADVH49B8249E AChemBlock

7-Bromo-6-azaindole-3-carboxylic acid

  • Cas Number : 1190314-78-3
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-6-azaindole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72416
Availability details for
Quantity
  • Availability
  • Add to Cart