ADVH49B824A0 AChemBlock

ethyl 6-(aminomethyl)nicotinate hydrochloride

  • Cas Number : 1189434-55-6
  • Molecular Weight : 216.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-aminomethyl-nicotinate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86527
Availability details for
Quantity
  • Availability
  • Add to Cart