ADVH49B824A2 AChemBlock

2-Morpholineacetic acid hydrochloride

  • Cas Number : 1187929-25-4
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-morpholineacetic-acid-hydrochloride-293704.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70394
Availability details for
Quantity
  • Availability
  • Add to Cart