ADVH49B824A4 AChemBlock

5H,6H,7H-pyrrolo[3,4-d]pyrimidine hydrochloride

  • Cas Number : 1187830-46-1
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5h-6h-7h-pyrrolo-3-4-d-pyrimidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82720
Availability details for
Quantity
  • Availability
  • Add to Cart