ADVH49B824A6 AChemBlock

tert-butyl (3-(bromomethyl)phenyl)carbamate

  • Cas Number : 118684-32-5
  • Molecular Weight : 286.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-bromomethyl-phenyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86567
Availability details for
Quantity
  • Availability
  • Add to Cart