ADVH49B824A8 AChemBlock

4,5-dimethylthiazole-2-thiol

  • Cas Number : 118090-07-6
  • Molecular Weight : 145.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-5-dimethylthiazole-2-thiol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71802
Availability details for
Quantity
  • Availability
  • Add to Cart