ADVH49B824AA AChemBlock

5-chloropyridine-2-carboximidamide hydrochloride

  • Cas Number : 1179360-48-5
  • Molecular Weight : 192.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-chloropyridine-2-carboximidamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82797
Availability details for
Quantity
  • Availability
  • Add to Cart