ADVH49B824AB AChemBlock

3-Aminopyrrolidin-2-one hydrochloride

  • Cas Number : 117879-49-9
  • Molecular Weight : 136.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-aminopyrrolidin-2-one-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82801
Availability details for
Quantity
  • Availability
  • Add to Cart