ADVH49B824AC AChemBlock

4-(pyridin-4-yl)-1H-pyrazole-3-carboxylic acid

  • Cas Number : 117784-27-7
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-pyridin-4-yl-1h-pyrazole-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82810
Availability details for
Quantity
  • Availability
  • Add to Cart