ADVH49B824AE AChemBlock

3-cyano-4-methoxybenzoic acid

  • Cas Number : 117738-82-6
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-cyano-4-methoxybenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82813
Availability details for
Quantity
  • Availability
  • Add to Cart