ADVH49B824B8 AChemBlock

4-fluoro-7-nitro-indoline

  • Cas Number : 1167056-12-3
  • Molecular Weight : 182.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-7-nitro-indoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82935
Availability details for
Quantity
  • Availability
  • Add to Cart